Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc4
Entrez Gene ID
362556
Gene Name
tetratricopeptide repeat domain 4
Gene Aliases
-
Chromosome Location
Chr.5: 126264758 - 126282429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238519.2 XP_006238581.1 8 - 9 1087 66

View Full Product Details