Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Awat1
Entrez Gene ID
679520
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases
Dgat2l3
Chromosome Location
Chr.X: 70408124 - 70414369 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109371.1 NP_001102841.1 1 - 2 101 90

View Full Product Details