Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syap1
Entrez Gene ID
302678
Gene Name
synapse associated protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 33528072 - 33562589 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004253.1 NP_001004253.1 5 - 6 770 72
XM_006256889.2 XP_006256951.1 5 - 6 783 72
GenBank mRNA BC079116.1 - 5 - 6 770 72
FQ219851.1 - 4 - 5 348 72

View Full Product Details