The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199256.2 | NP_954865.1 | 8 - 9 | 888 | 65 |
GenBank mRNA | BC094530.1 | - | 8 - 9 | 888 | 65 |
M96630.1 | - | 8 - 9 | 879 | 65 |