Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prok2
Entrez Gene ID
192206
Gene Name
prokineticin 2
Gene Aliases
Bv8
Chromosome Location
Chr.4: 132157556 - 132171153 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037541.1 NP_001032630.1 3 - 4 288 67
GenBank mRNA AY348322.1 - 3 - 4 288 67

View Full Product Details