Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo22
Entrez Gene ID
300724
Gene Name
F-box protein 22
Gene Aliases
-
Chromosome Location
Chr.8: 59770032 - 59786061 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037770.1 NP_001032859.1 4 - 5 483 68
GenBank mRNA BC107670.1 - 4 - 5 483 68

View Full Product Details