Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc2a10
Entrez Gene ID
366251
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 10
Gene Aliases
-
Chromosome Location
Chr.3: 162182156 - 162194610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108963.1 NP_001102433.1 1 - 2 118 71

View Full Product Details