Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco6c1
Entrez Gene ID
287006
Gene Name
solute carrier organic anion transporter family, member 6c1
Gene Aliases
GST-2
Chromosome Location
Chr.9: 104622219 - 104700657 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173338.1 NP_775460.1 - 898 135
XM_008767361.1 XP_008765583.1 - 780 135
GenBank mRNA AF326113.1 - - 898 135

View Full Product Details