The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024295.1 | NP_001019466.1 | 2 - 3 | 224 | 93 |
GenBank mRNA | BC083728.1 | - | 2 - 3 | 224 | 93 |