Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fundc1
Entrez Gene ID
363442
Gene Name
FUN14 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.X: 5549468 - 5566125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025027.1 NP_001020198.1 2 - 3 322 113
GenBank mRNA BC091353.1 - 2 - 3 322 113

View Full Product Details