Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx39a
Entrez Gene ID
89827
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A
Gene Aliases
Ddx39, Ddxl
Chromosome Location
Chr.19: 24846535 - 24854886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053563.2 NP_446015.2 3 - 4 636 73
XM_006255314.2 XP_006255376.1 3 - 4 536 73
GenBank mRNA AF063447.1 - 3 - 4 432 73
BC086328.1 - 3 - 4 636 73
FQ231639.1 - 3 - 4 541 73

View Full Product Details