The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106670.1 | NP_001100140.1 | 5 - 6 | 699 | 154 |
GenBank mRNA | FQ211202.1 | - | 5 - 6 | 778 | 154 |
FQ211244.1 | - | 5 - 6 | 778 | 154 |