The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108575.1 | NP_001102045.1 | 11 - 12 | 2257 | 86 |
XM_006233825.2 | XP_006233887.1 | 12 - 13 | 2311 | 86 | |
XM_006233826.2 | XP_006233888.1 | 12 - 13 | 2337 | 86 | |
XM_006233830.2 | XP_006233892.1 | 11 - 12 | 2260 | 86 | |
GenBank mRNA | BC168949.1 | - | 12 - 13 | 2256 | 86 |