Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brd3
Entrez Gene ID
362092
Gene Name
bromodomain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 5995204 - 6054470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108575.1 NP_001102045.1 1 - 2 108 59

View Full Product Details