Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sh3glb2
Entrez Gene ID
311848
Gene Name
SH3-domain GRB2-like endophilin B2
Gene Aliases
-
Chromosome Location
Chr.3: 8909231 - 8924055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009692.1 NP_001009692.1 2 - 3 260 90
XM_006233816.2 XP_006233878.1 2 - 3 283 90
XM_006233817.2 XP_006233879.1 2 - 3 283 90
XM_006233818.2 XP_006233880.1 2 - 3 282 90
XM_006233819.2 XP_006233881.1 2 - 3 282 90
XM_006233820.2 XP_006233882.1 2 - 3 282 90
XM_006233822.2 XP_006233884.1 2 - 3 282 90
XM_008761661.1 XP_008759883.1 2 - 3 283 90
XM_008761662.1 XP_008759884.1 2 - 3 282 90
GenBank mRNA BC087652.1 - 2 - 3 260 90

View Full Product Details