Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rxra
Entrez Gene ID
25271
Gene Name
retinoid X receptor alpha
Gene Aliases
-
Chromosome Location
Chr.3: 6272560 - 6295354 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012805.2 NP_036937.2 - 299 132
GenBank mRNA AM295012.1 - - 299 132
AM295013.1 - - 299 132
AM392401.1 - - 299 132
L06482.1 - - 466 132

View Full Product Details