Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ak8
Entrez Gene ID
311833
Gene Name
adenylate kinase 8
Gene Aliases
AK 8, RGD1303144
Chromosome Location
Chr.3: 7279429 - 7394509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004266.1 NP_001004266.1 6 - 7 500 86
XM_006233794.2 XP_006233856.1 7 - 8 536 86
GenBank mRNA BC079446.1 - 6 - 7 500 86

View Full Product Details