Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fzd7
Entrez Gene ID
301440
Gene Name
frizzled family receptor 7
Gene Aliases
-
Chromosome Location
Chr.9: 66057514 - 66153521 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767173.1 XP_008765395.1 7 - 8 1281 143

View Full Product Details