Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcnp
Entrez Gene ID
288165
Gene Name
PEST proteolytic signal containing nuclear protein
Gene Aliases
Ab2-416
Chromosome Location
Chr.11: 47055656 - 47070319 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047846.1 NP_001041311.1 5 - 6 484 91
XM_006248241.2 XP_006248303.1 4 - 5 457 91
XM_006248242.1 XP_006248304.1 4 - 5 455 91
XM_008768630.1 XP_008766852.1 4 - 5 516 91
XM_008768631.1 XP_008766853.1 4 - 5 513 91
GenBank mRNA AY325209.1 - 5 - 6 484 91

View Full Product Details