Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cck
Entrez Gene ID
25298
Gene Name
cholecystokinin
Gene Aliases
-
Chromosome Location
Chr.8: 130120524 - 130127295 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012829.2 NP_036961.1 1 - 2 107 51
XM_006244070.2 XP_006244132.1 1 - 2 293 51
GenBank mRNA M25942.1 - 1 - 2 30 51
X01032.1 - 1 - 2 107 51

View Full Product Details