The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106859.1 | NP_001100329.1 | 11 - 12 | 2149 | 87 |
XM_006243764.2 | XP_006243826.1 | 11 - 12 | 2148 | 87 |