Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Angptl6
Entrez Gene ID
298698
Gene Name
angiopoietin-like 6
Gene Aliases
-
Chromosome Location
Chr.8: 21895523 - 21902012 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106702.1 NP_001100172.1 4 - 5 1029 62
XM_008765921.1 XP_008764143.1 4 - 5 918 62
GenBank mRNA BC166828.1 - 4 - 5 992 62

View Full Product Details