Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dusp8
Entrez Gene ID
361679
Gene Name
dual specificity phosphatase 8
Gene Aliases
Hb5, M3/6
Chromosome Location
Chr.1: 215030429 - 215043111 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108510.1 NP_001101980.1 - 396 80

View Full Product Details