The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013962.1 | NP_001013984.1 | 2 - 3 | 209 | 119 |
GenBank mRNA | BC083676.1 | - | 2 - 3 | 209 | 119 |