Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fank1
Entrez Gene ID
309071
Gene Name
fibronectin type 3 and ankyrin repeat domains 1
Gene Aliases
-
Chromosome Location
Chr.1: 205842458 - 205950360 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008347.1 NP_001008348.1 9 - 10 999 83
GenBank mRNA BC082095.1 - 9 - 10 999 83

View Full Product Details