Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eef1g
Entrez Gene ID
293725
Gene Name
eukaryotic translation elongation factor 1 gamma
Gene Aliases
-
Chromosome Location
Chr.1: 225163391 - 225173997 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ212596.1 - 1 - 2 33 103
FQ219298.1 - 1 - 2 33 103
FQ235034.1 - 1 - 2 181 103

View Full Product Details