Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdcd1lg2
Entrez Gene ID
309304
Gene Name
programmed cell death 1 ligand 2
Gene Aliases
-
Chromosome Location
Chr.1: 247562202 - 247627193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107582.2 NP_001101052.2 2 - 3 288 136

View Full Product Details