The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014790.1 | NP_001014790.1 | 4 - 5 | 687 | 116 |
GenBank mRNA | AY936478.1 | - | 4 - 5 | 687 | 116 |
BC105631.1 | - | 4 - 5 | 687 | 116 |