Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ech1
Entrez Gene ID
64526
Gene Name
enoyl CoA hydratase 1, peroxisomal
Gene Aliases
Pxel
Chromosome Location
Chr.1: 87009798 - 87015996 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022594.1 NP_072116.1 4 - 5 481 74
GenBank mRNA BC060565.1 - 4 - 5 487 74
BC062226.1 - 4 - 5 487 74
FQ214121.1 - 4 - 5 483 74
FQ230435.1 - 4 - 5 494 74
U08976.1 - 4 - 5 481 74

View Full Product Details