Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cblc
Entrez Gene ID
292699
Gene Name
Cbl proto-oncogene C, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.1: 80699321 - 80716146 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034920.2 NP_001030092.1 5 - 6 940 99
GenBank mRNA BC105776.1 - 5 - 6 925 99

View Full Product Details