Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrp12
Entrez Gene ID
314941
Gene Name
low density lipoprotein receptor-related protein 12
Gene Aliases
-
Chromosome Location
Chr.7: 78859784 - 78930832 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134883.1 NP_001128355.1 2 - 3 223 79

View Full Product Details