The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004279.1 | NP_001004279.1 | 8 - 9 | 1044 | 69 |
GenBank mRNA | BC076386.1 | - | 8 - 9 | 1044 | 69 |
FQ214826.1 | - | 8 - 9 | 1047 | 69 |