The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008278.1 | NP_001008279.1 | 2 - 3 | 352 | 79 |
GenBank mRNA | BC086396.1 | - | 2 - 3 | 352 | 79 |