Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc16a5
Entrez Gene ID
690212
Gene Name
solute carrier family 16 (monocarboxylate transporter), member 5
Gene Aliases
-
Chromosome Location
Chr.10: 104019888 - 104035725 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109568.1 NP_001103038.1 1 - 2 304 90

View Full Product Details