Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac2
Entrez Gene ID
303692
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2
Gene Aliases
Siat7b
Chromosome Location
Chr.10: 105650866 - 105668579 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031652.1 NP_001026822.1 2 - 3 219 57
GenBank mRNA AJ634457.1 - 2 - 3 189 57
BC099812.1 - 2 - 3 219 57

View Full Product Details