Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo39
Entrez Gene ID
303287
Gene Name
F-box protein 39
Gene Aliases
-
Chromosome Location
Chr.10: 58874021 - 58878445 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039018.1 NP_001034107.1 2 - 3 1280 106
XM_006246727.2 XP_006246789.1 3 - 4 1537 106
XM_006246729.2 XP_006246791.1 3 - 4 1377 106
GenBank mRNA BC082089.1 - 3 - 4 1330 106
BC100079.1 - 2 - 3 1280 106

View Full Product Details