Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Emilin1
Entrez Gene ID
298845
Gene Name
elastin microfibril interfacer 1
Gene Aliases
-
Chromosome Location
Chr.6: 26821249 - 26829010 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106710.1 NP_001100180.1 3 - 4 989 87
XM_006239793.2 XP_006239855.1 3 - 4 1000 87
XM_008764511.1 XP_008762733.1 3 - 4 1732 87

View Full Product Details