The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007673.1 | NP_001007674.1 | 6 - 7 | 707 | 59 |
XM_006247126.1 | XP_006247188.1 | 6 - 7 | 745 | 59 | |
XM_006247127.2 | XP_006247189.1 | 6 - 7 | 1567 | 59 | |
GenBank mRNA | AF540876.1 | - | 6 - 7 | 606 | 59 |
BC081965.1 | - | 6 - 7 | 707 | 59 |