Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upk1b
Entrez Gene ID
303924
Gene Name
uroplakin 1B
Gene Aliases
-
Chromosome Location
Chr.11: 64522008 - 64553234 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024253.1 NP_001019424.1 7 - 8 791 157
XM_006248367.2 XP_006248429.1 7 - 8 998 157
XM_006248368.2 XP_006248430.1 7 - 8 838 157
GenBank mRNA BC093613.1 - 7 - 8 791 157

View Full Product Details