Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lin7b
Entrez Gene ID
60377
Gene Name
lin-7 homolog B (C. elegans)
Gene Aliases
Veli1a, Veli2
Chromosome Location
Chr.1: 101358313 - 101361056 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021758.1 NP_068526.1 2 - 3 153 78
GenBank mRNA AF090133.1 - 2 - 3 153 78

View Full Product Details