Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chsy1
Entrez Gene ID
292999
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
-
Chromosome Location
Chr.1: 127010587 - 127071570 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106268.1 NP_001099738.1 2 - 3 812 146

View Full Product Details