Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc88b
Entrez Gene ID
293698
Gene Name
coiled-coil domain containing 88B
Gene Aliases
RGD1311475
Chromosome Location
Chr.1: 222102436 - 222118479 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_215206.9 XP_215206.6 3 - 4 383 83

View Full Product Details