The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144737.2 | NP_653338.2 | 8 - 9 | 1348 | 109 |
XM_008769604.1 | XP_008767826.1 | 7 - 8 | 1612 | 109 | |
GenBank mRNA | AF458414.1 | - | 8 - 9 | 1315 | 107 |
BC070904.1 | - | 8 - 9 | 1348 | 109 |