Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
29231
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
Wdr39
Chromosome Location
Chr.3: 119671778 - 119677324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008766.1 NP_001008766.1 3 - 4 481 76
GenBank mRNA BC088474.1 - 3 - 4 481 76
FQ214368.1 - 3 - 4 504 76
FQ230934.1 - 3 - 4 495 76
FQ231672.1 - 3 - 4 474 76

View Full Product Details