The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106041.1 | NP_001099511.1 | 4 - 5 | 730 | 76 |
XM_006252041.1 | XP_006252103.1 | 4 - 5 | 642 | 76 |