Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March8
Entrez Gene ID
312656
Gene Name
membrane-associated ring finger (C3HC4) 8, E3 ubiquitin protein ligase
Gene Aliases
Mir
Chromosome Location
Chr.4: 148286854 - 148398147 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107882.1 NP_001101352.1 2 - 3 278 72

View Full Product Details