Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aggf1
Entrez Gene ID
310005
Gene Name
angiogenic factor with G patch and FHA domains 1
Gene Aliases
RGD1310888
Chromosome Location
Chr.2: 25068804 - 25095430 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006231820.2 XP_006231882.1 11 - 12 1935 152

View Full Product Details