The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001510.1 | NP_001001510.1 | 2 - 3 | 792 | 147 |
GenBank mRNA | AY495338.1 | - | 2 - 3 | 792 | 147 |
BC085842.1 | - | 2 - 3 | 770 | 147 |