Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt42
Entrez Gene ID
450231
Gene Name
keratin 42
Gene Aliases
Ka22
Chromosome Location
Chr.10: 88165667 - 88172910 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008816.1 NP_001008816.1 2 - 3 546 85

View Full Product Details