Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vat1
Entrez Gene ID
287721
Gene Name
vesicle amine transport 1
Gene Aliases
-
Chromosome Location
Chr.10: 89366898 - 89374516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033683.1 NP_001028855.1 5 - 6 1156 66
GenBank mRNA BC101882.1 - 5 - 6 1156 66

View Full Product Details